Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlh3
Entrez Gene ID
217716
Gene Name
mutL homolog 3 (E coli)
Gene Aliases
AV125803, BB126472
Chromosome Location
Chr.12: 85234466 - 85270648 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK029786.1 - 6 - 7 2281 70
AK132195.1 - 6 - 7 3689 70
AK162464.1 - 5 - 6 2356 70
BC003865.1 - 5 - 6 483 70
BC072599.1 - 6 - 7 3700 70
BC079861.1 - 6 - 7 3687 70

View Full Product Details