The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145523.3 | NP_663498.1 | 2 - 3 | 301 | 71 |
GenBank mRNA | AB088388.1 | - | 2 - 3 | 195 | 71 |
AF518325.1 | - | 2 - 3 | 285 | 71 | |
AK033740.1 | - | 2 - 3 | 334 | 71 | |
AK046347.1 | - | 2 - 3 | 343 | 71 | |
AK049317.1 | - | 2 - 3 | 302 | 71 | |
AK050540.1 | - | 2 - 3 | 343 | 71 | |
AK078915.1 | - | 2 - 3 | 302 | 71 | |
BC021450.1 | - | 2 - 3 | 295 | 71 |