Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnc4
Entrez Gene ID
99738
Gene Name
potassium voltage gated channel, Shaw-related subfamily, member 4
Gene Aliases
AI850292, KSHIIIC, Kcr2-4, Kv3.4
Chromosome Location
Chr.3: 107438296 - 107459952 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145922.2 NP_666034.1 1 - 2 703 94
GenBank mRNA AK158073.1 - 1 - 2 103 94
BC024837.1 - 1 - 2 703 94

View Full Product Details