The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145923.4 | NP_666035.2 | 6 - 7 | 989 | 75 |
GenBank mRNA | AK133845.1 | - | 7 - 8 | 1009 | 75 |
AK140108.1 | - | 6 - 7 | 989 | 75 | |
AK159944.1 | - | 6 - 7 | 915 | 75 | |
AK161061.1 | - | 6 - 7 | 915 | 75 | |
BC023848.1 | - | 2 - 3 | 285 | 75 | |
BC031198.1 | - | 6 - 7 | 958 | 75 | |
BC066137.1 | - | 6 - 7 | 968 | 75 | |
BC066160.1 | - | 6 - 7 | 968 | 75 |