The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145934.2 | NP_666046.1 | 4 - 5 | 426 | 76 |
NR_102313.1 | 4 - 5 | 426 | 76 | ||
GenBank mRNA | AK050131.1 | - | 4 - 5 | 406 | 76 |
AK050172.1 | - | 4 - 5 | 394 | 76 | |
BC026642.1 | - | 4 - 5 | 400 | 76 |