The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147778.3 | NP_680087.1 | 1 - 2 | 140 | 82 |
GenBank mRNA | AK007318.1 | - | 1 - 2 | 145 | 82 |
AK041634.1 | - | 1 - 2 | 141 | 82 | |
AK145955.1 | - | 1 - 2 | 168 | 82 | |
AK145999.1 | - | 1 - 2 | 170 | 82 | |
BC048815.1 | - | 1 - 2 | 151 | 82 |