The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_148935.2 | NP_683737.2 | 3 - 4 | 312 | 71 |
GenBank mRNA | AF323488.1 | - | 3 - 4 | 320 | 71 |
AY039039.1 | - | 3 - 4 | 358 | 71 | |
BC092242.1 | - | 3 - 4 | 301 | 71 | |
BC130222.1 | - | 3 - 4 | 313 | 71 |