Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxn4
Entrez Gene ID
116810
Gene Name
forkhead box N4
Gene Aliases
-
Chromosome Location
Chr.5: 114254164 - 114274598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_148935.2 NP_683737.2 5 - 6 556 87
GenBank mRNA AF323488.1 - 5 - 6 564 87
AK081735.1 - 3 - 4 261 87
AY039039.1 - 5 - 6 602 87
BC092242.1 - 5 - 6 545 87
BC130222.1 - 5 - 6 557 87

View Full Product Details