The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007896.3 | NP_031922.1 | 1 - 2 | 135 | 97 |
GenBank mRNA | AK154245.1 | - | 1 - 2 | 79 | 97 |
BC043333.1 | - | 1 - 2 | 99 | 97 | |
U51196.1 | - | 1 - 2 | 65 | 97 |