Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lig3
Entrez Gene ID
16882
Gene Name
ligase III, DNA, ATP-dependent
Gene Aliases
D11Wsu78e
Chromosome Location
Chr.11: 82781109 - 82804274 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291245.1 NP_001278174.1 8 - 9 1629 88
NM_001291246.1 NP_001278175.1 8 - 9 1626 88
NM_001291247.1 NP_001278176.1 8 - 9 1630 88
NM_010716.3 NP_034846.2 8 - 9 1633 88
GenBank mRNA AK031540.1 - 8 - 9 1570 88
AK143531.1 - 8 - 9 1499 88
AK150644.1 - 8 - 9 1538 88
AK156018.1 - 8 - 9 1548 88
BC026143.1 - 8 - 9 1601 88
BC049240.1 - 8 - 9 1536 88
BC083500.1 - 8 - 9 1532 88
U66057.1 - 8 - 9 1564 88
U66058.1 - 8 - 9 1564 88

View Full Product Details