Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lpin2
Entrez Gene ID
64898
Gene Name
lipin 2
Gene Aliases
2610511G02Rik, AI481352, AW742896
Chromosome Location
Chr.17: 71183528 - 71249818 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164885.1 NP_001158357.1 17 - 18 2556 138
NM_022882.4 NP_075020.2 17 - 18 2432 138
GenBank mRNA AF286723.1 - 17 - 18 2432 138
AK050138.1 - 3 - 4 348 138
AK165715.1 - 2 - 3 1988 138
BC022663.1 - 2 - 3 256 138
BC039698.1 - 17 - 18 2545 138

View Full Product Details