Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem63a
Entrez Gene ID
208795
Gene Name
transmembrane protein 63a
Gene Aliases
BC014795, Tmem64a
Chromosome Location
Chr.1: 180941225 - 180975106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144794.2 NP_659043.1 13 - 14 1276 63
GenBank mRNA AK049360.1 - 13 - 14 1271 63
AK144899.1 - 13 - 14 1329 63
AK170559.1 - 13 - 14 1292 63
AK173027.1 - 13 - 14 1269 63
BC014795.1 - 13 - 14 1249 63
BC019442.1 - 13 - 14 1276 63

View Full Product Details