Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Acsl6
Entrez Gene ID
216739
Gene Name
acyl-CoA synthetase long-chain family member 6
Gene Aliases
A330035H04Rik, AW050338, Facl6, LACS, Lacsl, mKIAA0837
Chromosome Location
Chr.11: 54303824 - 54364756 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033597.1 NP_001028769.1 3 - 4 464 96
NM_001033598.1 NP_001028770.1 3 - 4 433 96
NM_001033599.1 NP_001028771.1 3 - 4 639 96
NM_144823.4 NP_659072.3 3 - 4 464 96
GenBank mRNA AB073974.1 - 2 - 3 308 96
AK032479.1 - 4 - 5 723 96
AK039380.1 - 3 - 4 550 96
AK147283.1 - 3 - 4 434 96
AK147421.1 - 3 - 4 465 96
AY167035.1 - 3 - 4 434 96
AY786360.1 - 5 - 6 982 96
AY786361.1 - 3 - 4 414 96
AY786362.1 - 3 - 4 606 96
AY786363.1 - 3 - 4 459 96
BC016114.1 - 3 - 4 390 96
BC022959.1 - 3 - 4 464 96

View Full Product Details