Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Utp6
Entrez Gene ID
216987
Gene Name
UTP6, small subunit (SSU) processome component, homolog (yeast)
Gene Aliases
4732497O03Rik, HCA66
Chromosome Location
Chr.11: 79932320 - 79962426 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144826.3 NP_659075.1 11 - 12 1044 68
GenBank mRNA AF334387.1 - 11 - 12 1371 68
AK029151.1 - 11 - 12 1045 68
AK084976.1 - 11 - 12 1037 68
AK164010.1 - 11 - 12 1050 68
AK169785.1 - 11 - 12 1051 68
BC018250.1 - 11 - 12 1008 68
BC023920.1 - 11 - 12 1038 68
BC024850.1 - 11 - 12 1039 68

View Full Product Details