The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144837.3 | NP_659086.2 | 14 - 15 | 5989 | 75 |
GenBank mRNA | AK129244.1 | - | 2 - 3 | 4426 | 75 |
AK134564.1 | - | 2 - 3 | 2432 | 75 | |
BC018507.1 | - | 1 - 2 | 84 | 75 | |
BC026613.1 | - | 2 - 3 | 515 | 75 |