The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144843.4 | NP_659092.1 | 6 - 7 | 861 | 73 |
GenBank mRNA | AK076218.1 | - | 6 - 7 | 838 | 73 |
AK154859.1 | - | 6 - 7 | 781 | 73 | |
BC020019.1 | - | 6 - 7 | 795 | 73 |