The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144861.2 | NP_659110.1 | 6 - 7 | 930 | 65 |
GenBank mRNA | AK137068.1 | - | 5 - 6 | 1583 | 65 |
AK220475.1 | - | 6 - 7 | 930 | 65 | |
BC021395.1 | - | 6 - 7 | 882 | 65 | |
BC023084.1 | - | 6 - 7 | 867 | 65 |