Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slmo1
Entrez Gene ID
225655
Gene Name
slowmo homolog 1 (Drosophila)
Gene Aliases
A230074A19Rik, BC019561
Chromosome Location
Chr.18: 67464835 - 67480581 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144867.2 NP_659116.1 5 - 6 547 71
GenBank mRNA AK143220.1 - 5 - 6 531 71
BC019561.1 - 5 - 6 496 71
BY741062.1 - 5 - 6 548 71

View Full Product Details