The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144872.1 | NP_659121.1 | 4 - 5 | 806 | 71 |
GenBank mRNA | BC022146.1 | - | 4 - 5 | 806 | 71 |
BC023878.1 | - | 4 - 5 | 829 | 71 | |
BC034112.1 | - | 4 - 5 | 604 | 71 |