The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206382.1 | NP_001193311.1 | 6 - 7 | 836 | 84 |
NM_001206383.1 | NP_001193312.1 | 4 - 5 | 570 | 84 | |
NM_001206385.1 | NP_001193314.1 | 5 - 6 | 745 | 84 | |
NM_144888.2 | NP_659137.1 | 5 - 6 | 973 | 84 | |
GenBank mRNA | AK028421.1 | - | 5 - 6 | 744 | 84 |
AK085038.1 | - | 5 - 6 | 932 | 84 | |
AK153748.1 | - | 6 - 7 | 838 | 84 | |
AX775773.1 | - | 5 - 6 | 722 | 84 | |
BC020006.1 | - | 5 - 6 | 705 | 84 | |
BC025825.1 | - | 5 - 6 | 898 | 84 | |
BC031352.1 | - | 5 - 6 | 882 | 84 | |
BC037391.1 | - | 4 - 5 | 684 | 84 | |
DQ167127.1 | - | 4 - 5 | 622 | 84 | |
DQ174271.1 | - | 4 - 5 | 622 | 84 |