Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmf1
Entrez Gene ID
228769
Gene Name
proteasome (prosome, macropain) inhibitor subunit 1
Gene Aliases
AW048666, BC012260, PI31
Chromosome Location
Chr.2: 151716062 - 151744188 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK031363.1 - 4 - 5 708 90
AK045891.1 - 4 - 5 691 90
AK053924.1 - 4 - 5 709 90
AK078867.1 - 4 - 5 667 90
BC012260.1 - 4 - 5 668 90
BC128314.1 - 3 - 4 453 90
BC131928.1 - 4 - 5 572 90
BC131930.1 - 4 - 5 572 90

View Full Product Details