Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bpifb5
Entrez Gene ID
228802
Gene Name
BPI fold containing family B, member 5
Gene Aliases
Lplunc5
Chromosome Location
Chr.2: 154223742 - 154240902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144890.2 NP_659139.2 5 - 6 693 74
GenBank mRNA AK142982.1 - 5 - 6 694 74
BC018465.1 - 5 - 6 655 74

View Full Product Details