The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144892.1 | NP_659141.1 | 6 - 7 | 948 | 77 |
GenBank mRNA | AK049242.1 | - | 3 - 4 | 352 | 77 |
BC017152.1 | - | 6 - 7 | 948 | 77 |