Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gckr
Entrez Gene ID
231103
Gene Name
glucokinase regulatory protein
Gene Aliases
GKRP
Chromosome Location
Chr.5: 31297254 - 31327302 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144909.1 NP_659158.1 18 - 19 1641 85
GenBank mRNA AK149377.1 - 16 - 17 3648 85
BC012412.1 - 18 - 19 1641 85

View Full Product Details