The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285511.1 | NP_001272440.1 | 2 - 3 | 307 | 109 |
NM_144910.2 | NP_659159.1 | 2 - 3 | 175 | 109 | |
NM_178854.4 | NP_849185.2 | 2 - 3 | 185 | 109 | |
GenBank mRNA | AK151728.1 | - | 2 - 3 | 310 | 109 |
BC018506.1 | - | 2 - 3 | 175 | 109 | |
CF173938.1 | - | 2 - 3 | 109 |