The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163461.2 | NP_001156933.1 | 6 - 7 | 578 | 74 |
NM_001163462.2 | NP_001156934.1 | 6 - 7 | 635 | 74 | |
NM_001289569.1 | NP_001276498.1 | 6 - 7 | 592 | 74 | |
NM_001289570.1 | NP_001276499.1 | 6 - 7 | 578 | 74 | |
NM_144911.3 | NP_659160.2 | 6 - 7 | 578 | 74 | |
GenBank mRNA | AK034418.1 | - | 6 - 7 | 594 | 74 |
AK049889.1 | - | 6 - 7 | 635 | 74 | |
AK145830.1 | - | 6 - 7 | 553 | 74 | |
AK161901.1 | - | 6 - 7 | 559 | 74 | |
AK166574.1 | - | 6 - 7 | 501 | 74 | |
BC021895.1 | - | 6 - 7 | 539 | 74 |