Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rad9b
Entrez Gene ID
231724
Gene Name
RAD9 homolog B
Gene Aliases
A630082N15Rik, BC021784
Chromosome Location
Chr.5: 122323223 - 122354272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144912.3 NP_659161.2 3 - 4 283 113
GenBank mRNA AK136504.1 - 3 - 4 319 113
AY297460.1 - 3 - 4 283 113
BC021784.1 - 3 - 4 283 113

View Full Product Details