Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smyd5
Entrez Gene ID
232187
Gene Name
SET and MYND domain containing 5
Gene Aliases
AW536703, NN8-4AG, Rai15, Rrg1
Chromosome Location
Chr.6: 85431976 - 85446429 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144918.2 NP_659167.2 6 - 7 665 63
GenBank mRNA AK158267.1 - 6 - 7 646 63
AK162926.1 - 6 - 7 589 63
BC016525.1 - 6 - 7 652 63

View Full Product Details