The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145532.3 | NP_663507.1 | 1 - 2 | 141 | 60 |
GenBank mRNA | AK078971.1 | - | 1 - 2 | 142 | 60 |
AK163867.1 | - | 1 - 2 | 142 | 60 | |
BC012256.1 | - | 1 - 2 | 176 | 60 |