Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Yipf1
Entrez Gene ID
230584
Gene Name
Yip1 domain family, member 1
Gene Aliases
C030002N13Rik
Chromosome Location
Chr.4: 107314254 - 107359823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205156.1 NP_001192085.1 7 - 8 898 71
NM_145550.3 NP_663525.1 8 - 9 979 71
GenBank mRNA AI303400.1 - 2 - 3 71
AK048749.1 - 2 - 3 1750 71
AK081205.1 - 8 - 9 966 71
AK157536.1 - 8 - 9 966 71
BC009080.1 - 8 - 9 911 71

View Full Product Details