The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003898.3 | NP_001003898.1 | 1 - 2 | 315 | 134 |
NM_001003899.2 | NP_001003899.1 | 1 - 2 | 315 | 134 | |
NM_001008545.2 | NP_001008545.1 | 1 - 2 | 315 | 134 | |
NM_001008546.2 | NP_001008546.1 | 1 - 2 | 315 | 134 | |
NM_145556.4 | NP_663531.1 | 1 - 2 | 315 | 134 | |
NR_027864.1 | 1 - 2 | 315 | 134 | ||
GenBank mRNA | AK032199.1 | - | 1 - 2 | 85 | 134 |
AK045495.1 | - | 1 - 2 | 85 | 134 | |
AK083312.1 | - | 1 - 2 | 89 | 134 | |
AK146002.1 | - | 1 - 2 | 85 | 134 | |
AK146997.1 | - | 1 - 2 | 85 | 134 | |
BC012873.1 | - | 1 - 2 | 73 | 134 | |
BC027105.1 | - | 1 - 2 | 77 | 134 | |
BC027772.1 | - | 1 - 2 | 75 | 134 | |
BC029728.1 | - | 1 - 2 | 76 | 134 | |
BC031126.1 | - | 1 - 2 | 77 | 134 | |
BC033475.1 | - | 1 - 2 | 73 | 134 | |
CD548243.1 | - | 1 - 2 | 315 | 134 |