The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003898.3 | NP_001003898.1 | 3 - 4 | 733 | 160 |
NM_001003899.2 | NP_001003899.1 | 3 - 4 | 733 | 160 | |
NM_001008545.2 | NP_001008545.1 | 3 - 4 | 733 | 160 | |
NM_001008546.2 | NP_001008546.1 | 3 - 4 | 733 | 160 | |
NM_145556.4 | NP_663531.1 | 3 - 4 | 733 | 160 | |
NR_027864.1 | 3 - 4 | 733 | 160 | ||
GenBank mRNA | AK032199.1 | - | 3 - 4 | 503 | 160 |
AK045495.1 | - | 3 - 4 | 503 | 160 | |
AK083312.1 | - | 3 - 4 | 507 | 160 | |
AK133207.1 | - | 3 - 4 | 510 | 160 | |
AK146002.1 | - | 3 - 4 | 503 | 160 | |
AK146997.1 | - | 4 - 5 | 412 | 160 | |
AY319513.1 | - | 2 - 3 | 403 | 160 | |
AY319514.1 | - | 2 - 3 | 403 | 160 | |
AY319515.1 | - | 2 - 3 | 403 | 160 | |
AY319516.1 | - | 2 - 3 | 403 | 160 | |
BC012873.1 | - | 3 - 4 | 491 | 160 | |
BC025544.1 | - | 3 - 4 | 504 | 160 | |
BC027105.1 | - | 3 - 4 | 495 | 160 | |
BC027772.1 | - | 3 - 4 | 493 | 160 | |
BC029728.1 | - | 3 - 4 | 494 | 160 | |
BC031126.1 | - | 3 - 4 | 495 | 160 | |
BC033475.1 | - | 3 - 4 | 491 | 160 | |
GQ428775.1 | - | 2 - 3 | 403 | 160 | |
GQ428776.1 | - | 2 - 3 | 403 | 160 | |
GQ428777.1 | - | 2 - 3 | 403 | 160 | |
GU250735.1 | - | 2 - 3 | 403 | 160 |