The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181323.2 | NP_851840.1 | 4 - 5 | 793 | 58 |
GenBank mRNA | AK035550.1 | - | 4 - 5 | 644 | 58 |
AK081976.1 | - | 4 - 5 | 793 | 58 | |
BC016523.1 | - | 4 - 5 | 621 | 58 |