The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145561.2 | NP_663536.1 | 7 - 8 | 716 | 73 |
GenBank mRNA | AB053953.1 | - | 7 - 8 | 750 | 73 |
AF448809.1 | - | 7 - 8 | 695 | 73 | |
AF539752.1 | - | 2 - 3 | 289 | 73 | |
BC020151.1 | - | 7 - 8 | 716 | 73 |