The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145561.2 | NP_663536.1 | 9 - 10 | 1128 | 82 |
GenBank mRNA | AB053953.1 | - | 9 - 10 | 1162 | 82 |
AF448809.1 | - | 9 - 10 | 1107 | 82 | |
AF539752.1 | - | 4 - 5 | 701 | 82 | |
BC020151.1 | - | 9 - 10 | 1128 | 82 |