Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pgap2
Entrez Gene ID
233575
Gene Name
post-GPI attachment to proteins 2
Gene Aliases
1810006G21Rik, C76649, Frag1
Chromosome Location
Chr.7: 102210242 - 102238564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291358.1 NP_001278287.1 4 - 5 795 74
NM_145583.3 NP_663558.1 4 - 5 661 74
NR_111937.1 3 - 4 398 74
GenBank mRNA AK134287.1 - 4 - 5 678 74
AK146442.1 - 4 - 5 619 74
AK152420.1 - 3 - 4 602 74
AK152931.1 - 3 - 4 333 74
AK159933.1 - 4 - 5 636 74
AK159950.1 - 4 - 5 634 74
BC004794.1 - 4 - 5 615 74

View Full Product Details