Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ppan
- Entrez Gene ID
-
235036
- Gene Name
- peter pan homolog (Drosophila)
- Gene Aliases
-
A230087P06Rik, SSF1
- Chromosome Location
-
Chr.9: 20888175 - 20892179 on Build GRCm38
- Gene Symbol
- B130055M24Rik
- Entrez Gene ID
-
654803
- Gene Name
- RIKEN cDNA B130055M24 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 127448654 - 127461238 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK156871.1 |
- |
- |
563 |
60 |
View Full Product Details