Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lace1
Entrez Gene ID
215951
Gene Name
lactation elevated 1
Gene Aliases
CG8520
Chromosome Location
Chr.10: 42312581 - 42478584 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145743.2 NP_665686.2 1 - 2 299 61
GenBank mRNA AF520417.1 - 1 - 2 148 61
AK044595.1 - 1 - 2 175 61
AK170996.1 - 1 - 2 150 61
BC089595.1 - 1 - 2 149 61

View Full Product Details