The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145949.2 | NP_666061.3 | 9 - 10 | 952 | 66 |
GenBank mRNA | AK082385.1 | - | 8 - 9 | 1143 | 66 |
BC026393.1 | - | 9 - 10 | 952 | 66 | |
EF137182.1 | - | 9 - 10 | 952 | 66 |