The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163622.1 | NP_001157094.1 | 10 - 11 | 2006 | 68 |
NM_001163623.1 | NP_001157095.1 | 9 - 10 | 1470 | 68 | |
NM_001163624.1 | NP_001157096.1 | 9 - 10 | 1338 | 68 | |
NM_145984.3 | NP_666096.3 | 10 - 11 | 1570 | 68 | |
GenBank mRNA | AK028877.1 | - | 10 - 11 | 1964 | 68 |
AK050878.1 | - | 9 - 10 | 1332 | 68 | |
AK083070.1 | - | 10 - 11 | 1511 | 68 | |
AK129142.1 | - | 4 - 5 | 1424 | 68 | |
BC004612.1 | - | 9 - 10 | 1424 | 68 |