Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem82
Entrez Gene ID
213989
Gene Name
transmembrane protein 82
Gene Aliases
-
Chromosome Location
Chr.4: 141614229 - 141618633 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145987.2 NP_666099.2 5 - 6 1593 82
GenBank mRNA AK085940.1 - - 1569 82
AK162322.1 - 4 - 5 2013 82
BC025833.1 - 5 - 6 994 82

View Full Product Details