The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289711.1 | NP_001276640.1 | 8 - 9 | 1059 | 68 |
NM_001289712.1 | NP_001276641.1 | 8 - 9 | 1059 | 68 | |
NM_145993.5 | NP_666105.2 | 8 - 9 | 1059 | 68 | |
NR_110362.1 | 8 - 9 | 1059 | 68 | ||
GenBank mRNA | AK029115.1 | - | 8 - 9 | 1004 | 68 |
AK036510.1 | - | 8 - 9 | 1012 | 68 | |
AK132012.1 | - | 8 - 9 | 1012 | 68 | |
AK141810.1 | - | 7 - 8 | 2193 | 68 | |
AK145560.1 | - | 8 - 9 | 1001 | 68 | |
AK146632.1 | - | 8 - 9 | 1015 | 68 | |
AK156236.1 | - | 8 - 9 | 1014 | 68 | |
AK184628.1 | - | 1 - 2 | 50 | 68 | |
AY237000.1 | - | 8 - 9 | 992 | 68 | |
AY237001.1 | - | 8 - 9 | 992 | 68 | |
AY237002.1 | - | 8 - 9 | 992 | 68 | |
BC023933.1 | - | 8 - 9 | 1000 | 68 | |
BC030864.1 | - | 8 - 9 | 962 | 68 |