Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bud13
Entrez Gene ID
215051
Gene Name
BUD13 homolog (yeast)
Gene Aliases
AV305342, D030060M11Rik
Chromosome Location
Chr.9: 46283012 - 46298783 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146000.2 NP_666112.1 6 - 7 1438 73
GenBank mRNA AK037134.1 - 6 - 7 1435 73
AK038126.1 - 6 - 7 1443 73
AK051047.1 - 6 - 7 1443 73
AK077744.1 - 6 - 7 1442 73
BC025490.1 - 6 - 7 1439 73

View Full Product Details