The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146028.4 | NP_666140.1 | 1 - 2 | 569 | 80 |
GenBank mRNA | AJ608761.1 | - | 1 - 2 | 382 | 80 |
AK078132.1 | - | 1 - 2 | 570 | 80 | |
AK079357.1 | - | 1 - 2 | 427 | 80 | |
AK144433.1 | - | 1 - 2 | 425 | 80 | |
BC024864.1 | - | 1 - 2 | 475 | 80 | |
BC029794.1 | - | 1 - 2 | 416 | 80 |