Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt79
Entrez Gene ID
223917
Gene Name
keratin 79
Gene Aliases
BC031593
Chromosome Location
Chr.15: 101929332 - 101940324 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146063.1 NP_666175.1 4 - 5 957 75
GenBank mRNA BC019155.1 - 4 - 5 896 75
BC031593.1 - 4 - 5 957 75

View Full Product Details