Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cpped1
- Entrez Gene ID
-
223978
- Gene Name
- calcineurin-like phosphoesterase domain containing 1
- Gene Aliases
-
C530044N13Rik
- Chromosome Location
-
Chr.16: 11803684 - 11909475 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC034347.1 |
- |
2 - 3 |
210 |
69 |
View Full Product Details