The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146084.1 | NP_666196.1 | 17 - 18 | 2580 | 74 |
GenBank mRNA | AK029766.1 | - | 18 - 19 | 2696 | 74 |
BC029030.1 | - | 1 - 2 | 228 | 74 | |
BC031465.1 | - | 17 - 18 | 2580 | 74 |