Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Afap1l2
Entrez Gene ID
226250
Gene Name
actin filament associated protein 1-like 2
Gene Aliases
AU041783, C86904, mKIAA1914
Chromosome Location
Chr.19: 56912354 - 57008590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177796.1 NP_001171267.1 3 - 4 609 75
NM_001177797.1 NP_001171268.1 2 - 3 555 75
NM_146102.2 NP_666214.1 4 - 5 661 75
GenBank mRNA AK048649.1 - 4 - 5 315 75
AK053222.1 - 6 - 7 911 75
AK087449.1 - 3 - 4 245 75
AK087809.1 - 2 - 3 189 75
AK220430.1 - 2 - 3 178 75
BC031515.1 - 3 - 4 211 75

View Full Product Details