The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146152.3 | NP_666264.1 | 15 - 16 | 3028 | 95 |
GenBank mRNA | AK052257.1 | - | 15 - 16 | 3029 | 95 |
AK129200.1 | - | 15 - 16 | 2990 | 95 | |
AK149978.1 | - | 15 - 16 | 3011 | 95 | |
BC031823.1 | - | 15 - 16 | 2427 | 95 |