The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163472.1 | NP_001156944.1 | 2 - 3 | 437 | 70 |
NM_019823.4 | NP_062797.3 | 2 - 3 | 431 | 70 | |
GenBank mRNA | AF221525.1 | - | 1 - 2 | 227 | 70 |
AK090296.1 | - | 1 - 2 | 97 | 70 | |
BC006853.1 | - | 2 - 3 | 437 | 70 | |
BC016256.1 | - | 2 - 3 | 339 | 70 | |
BX520002.1 | - | 2 - 3 | 398 | 70 |